accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOCl

CAS RN 3441-03-0

IUPAC Name: methyl 3-(carbonochloridoyl)benzoate
Synonyms: methyl 3-(carbonochloridoyl)benzoate
Molecular Weight (g/mol): 198.6
Molecular Formula: C9H7ClO3
InChi Key: UTGGIQHJOAPIPD-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC=C1)C(Cl)=O
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)