accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3443-45-6

IUPAC Name: 4-(pyren-1-yl)butanoic acid
Synonyms: 4-(pyren-1-yl)butanoic acid
Molecular Weight (g/mol): 288.35
Molecular Formula: C20H16O2
InChi Key: QXYRRCOJHNZVDJ-UHFFFAOYSA-N
SMILES: OC(=O)CCCC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)