accessibility menu, dialog, popup

UserName

Viewing profile as user:
CrH3CCH3OOH3CCH3OOH3CCH3OO

CAS RN 3444-17-5

IUPAC Name: chromium(3+) tris(2-ethylhexanoate)
Synonyms: chromium(III) 2-ethylhexanoate
Molecular Weight (g/mol): 481.61
Molecular Formula: C24H45CrO6
InChi Key: WBKDDMYJLXVBNI-UHFFFAOYNA-K
SMILES: [Cr+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)