CAS RN 3445-11-2
IUPAC Name:
1-(2-hydroxyethyl)pyrrolidin-2-one
Synonyms:
1-(2-hydroxyethyl)pyrrolidin-2-one
Molecular Weight (g/mol):
129.16
Molecular Formula:
C6H11NO2
InChi Key:
WDQFELCEOPFLCZ-UHFFFAOYSA-N
SMILES:
OCCN1CCCC1=O
1
–
1
of
1
results
1
1-(2-Hydroxyethyl)-2-pyrrolidone 98.0+%, TCI America™
CAS: 3445-11-2 Molecular Formula: C6H11NO2 Molecular Weight (g/mol): 129.16 MDL Number: MFCD00014103 InChI Key: WDQFELCEOPFLCZ-UHFFFAOYSA-N PubChem CID: 76980 IUPAC Name: 1-(2-hydroxyethyl)pyrrolidin-2-one SMILES: C1CC(=O)N(C1)CCO