accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNN

CAS RN 3445-84-9

IUPAC Name: N,N-bis(2-cyanoethyl)formamide
Synonyms: N,N-bis(2-cyanoethyl)formamide
Molecular Weight (g/mol): 151.17
Molecular Formula: C7H9N3O
InChi Key: MYRFNYCEQURXPT-UHFFFAOYSA-N
SMILES: O=CN(CCC#N)CCC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results