CAS RN 34451-19-9
IUPAC Name:
butyl 2-hydroxypropanoate
Synonyms:
butyl lactate N-butyl lactate
Molecular Weight (g/mol):
146.19
Molecular Formula:
C7H14O3
InChi Key:
MRABAEUHTLLEML-UHFFFAOYNA-N
SMILES:
CCCCOC(=O)C(C)O
1
–
2
of
2
results
1
Promotions Available
n-Butyl L-lactate, 97+%
n-Butyl L-lactate, 97+%
CAS: 34451-19-9 Molecular Formula: C7H14O3 Molecular Weight (g/mol): 146.19 MDL Number: MFCD00004519,MFCD00066454 InChI Key: MRABAEUHTLLEML-UHFFFAOYNA-N PubChem CID: 6999977 IUPAC Name: butyl (2S)-2-hydroxypropanoate SMILES: CCCCOC(=O)C(C)O
2
Sigma Aldrich Bis(2,4,6-Trimethylphenyl)Phosphorus Chloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.