accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOClF

CAS RN 345-18-6

IUPAC Name: 4-chloro-1-fluoro-2-nitrobenzene
Synonyms: 4-chloro-1-fluoro-2-nitrobenzene
Molecular Weight (g/mol): 175.54
Molecular Formula: C6H3ClFNO2
InChi Key: DIAWBHLTWNWYGR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=CC=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)