accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 345-35-7

IUPAC Name: 1-(chloromethyl)-2-fluorobenzene
Synonyms: O-fluorobenzyl chloride
Molecular Weight (g/mol): 144.57
Molecular Formula: C7H6ClF
InChi Key: MOBRMRJUKNQBMY-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1CCl
Boiling Point 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)