accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOF

CAS RN 345-92-6

IUPAC Name: bis(4-fluorophenyl)methanone
Synonyms: 4,4'-difluorobenzophenone
Molecular Weight (g/mol): 218.2
Molecular Formula: C13H8F2O
InChi Key: LSQARZALBDFYQZ-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)C1=CC=C(F)C=C1
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (9)
Percent Purity 
  • (3)
  • (3)
  • (5)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (3)
  • (6)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (9)
  • (3)
  • (3)
  • (5)
  • (3)
  • (3)
  • (6)
  • (1)