accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH

CAS RN 3452-09-3

IUPAC Name: non-1-yne
Synonyms: 1-nonyne
Molecular Weight (g/mol): 124.23
Molecular Formula: C9H16
InChi Key: OSSQSXOTMIGBCF-UHFFFAOYSA-N
SMILES: CCCCCCCC#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results