accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3453-00-7

IUPAC Name: diethyl (2-oxo-2-phenylethyl)phosphonate
Synonyms: diethyl 2-oxo-2-phenylethylphosphonate
Molecular Weight (g/mol): 256.24
Molecular Formula: C12H17O4P
InChi Key: HPEVTTNSIPGLEL-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC(=O)C1=CC=CC=C1)OCC
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)