accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NClF

CAS RN 34552-15-3

IUPAC Name: 2-chloro-3-fluoro-5-methylpyridine
Synonyms: 2-chloro-3-fluoro-5-methylpyridine
Molecular Weight (g/mol): 145.56
Molecular Formula: C6H5ClFN
InChi Key: XZJURWKNRAGMCG-UHFFFAOYSA-N
SMILES: CC1=CC(F)=C(Cl)N=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)