accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3457-46-3

IUPAC Name: bis(4-chlorophenyl)ethane-1,2-dione
Synonyms: bis(4-chlorophenyl)ethane-1,2-dione
Molecular Weight (g/mol): 279.12
Molecular Formula: C14H8Cl2O2
InChi Key: XMAWUPHYEABFDR-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(=O)C(=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results