accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3

CAS RN 3457-48-5

IUPAC Name: bis(4-methylphenyl)ethane-1,2-dione
Synonyms: bis(4-methylphenyl)ethane-1,2-dione
Molecular Weight (g/mol): 238.29
Molecular Formula: C16H14O2
InChi Key: BCWCEHMHCDCJAD-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)C(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)