accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOCaH3CH3CCH3CH3

CAS RN 345909-31-1

IUPAC Name: calcium bis((2Z)-4-oxopent-2-en-2-olate)
Synonyms: calcium bis((2Z)-4-oxopent-2-en-2-olate)
Molecular Weight (g/mol): 238.3
Molecular Formula: C10H14CaO4
InChi Key: QAZYYQMPRQKMAC-FDGPNNRMSA-L
SMILES: [Ca++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results