CAS RN 34601-54-2
CAS RN 34601-54-2
IUPAC Name:
1,3,6,8-tetra-tert-butyl-9H-carbazole
Synonyms:
1,3,6,8-tetra-tert-butyl-9H-carbazole
Molecular Weight (g/mol):
391.64
Molecular Formula:
C28H41N
InChi Key:
OVSGNPWPCZRNKI-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC2=C(NC3=C2C=C(C=C3C(C)(C)C)C(C)(C)C)C(=C1)C(C)(C)C
1
–
1
of
1
results
1
1,3,6,8-Tetra-tert-butylcarbazole 98.0+%, TCI America™
CAS: 34601-54-2 Molecular Formula: C28H41N Molecular Weight (g/mol): 391.643 MDL Number: MFCD00022212 InChI Key: OVSGNPWPCZRNKI-UHFFFAOYSA-N PubChem CID: 545459 IUPAC Name: 1,3,6,8-tetratert-butyl-9H-carbazole SMILES: CC(C)(C)C1=CC(=C2C(=C1)C3=CC(=CC(=C3N2)C(C)(C)C)C(C)(C)C)C(C)(C)C