accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3Cl

CAS RN 346656-42-6

IUPAC Name: 2-(2-chlorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
Synonyms: 2-(2-chlorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
Molecular Weight (g/mol): 224.49
Molecular Formula: C11H14BClO2
InChi Key: HTKUDRCYEPYTOR-UHFFFAOYSA-N
SMILES: CC1(C)COB(OC1)C1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results