accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 346695-75-8

IUPAC Name: 2-chloro-4-nitro-N,N-dipropylbenzamide
Synonyms: 2-chloro-4-nitro-N,N-dipropylbenzamide
Molecular Weight (g/mol): 284.74
Molecular Formula: C13H17ClN2O3
InChi Key: WIYZJWRFZCMKTB-UHFFFAOYSA-N
SMILES: CCCN(CCC)C(=O)C1=C(Cl)C=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results