accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3HNCH3O

CAS RN 3467-59-2

IUPAC Name: N-(2,5-dimethoxyphenyl)acetamide
Synonyms: N-(2,5-dimethoxyphenyl)acetamide
Molecular Weight (g/mol): 195.22
Molecular Formula: C10H13NO3
InChi Key: HOZRMSVNIKYCMB-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(NC(C)=O)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results