accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3468-17-5

IUPAC Name: 1-(1H-indol-6-yl)methanamine
Synonyms: 1-(1H-indol-6-yl)methanamine
Molecular Weight (g/mol): 146.19
Molecular Formula: C9H10N2
InChi Key: FURRUNQWZZOXOT-UHFFFAOYSA-N
SMILES: NCC1=CC=C2C=CNC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)