accessibility menu, dialog, popup

UserName

Viewing profile as user:
INHN

CAS RN 3469-69-0

IUPAC Name: 4-iodo-1H-pyrazole
Synonyms: 4-iodopyrazole
Molecular Weight (g/mol): 193.98
Molecular Formula: C3H3IN2
InChi Key: LLNQWPTUJJYTTE-UHFFFAOYSA-N
SMILES: IC1=CNN=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (4)
Quantity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (3)
  • (5)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (3)
  • (4)