accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHO

CAS RN 3470-50-6

IUPAC Name: 6-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms: 6-hydroxy-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: FNSQPQKPPGALFA-UHFFFAOYSA-N
SMILES: OC1=CC2=C(C=C1)C(=O)CCC2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results