accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NO

CAS RN 3470-53-9

IUPAC Name: 6-amino-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms: 6-amino-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: BEVVUJBVEXJGKM-UHFFFAOYSA-N
SMILES: NC1=CC=C2C(=O)CCCC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)