accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3H2N

CAS RN 34701-33-2

IUPAC Name: 3-methylbutan-2-amine
Synonyms: 3-methyl-2-butanamine
Molecular Weight (g/mol): 87.17
Molecular Formula: C5H13N
InChi Key: JOZZAIIGWFLONA-UHFFFAOYNA-N
SMILES: CC(C)C(C)N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results