accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34732-09-7

IUPAC Name: 2,3,4-trichlorobenzene-1-sulfonyl chloride
Synonyms: 2,3,4-trichlorobenzenesulfonyl chloride
Molecular Weight (g/mol): 279.94
Molecular Formula: C6H2Cl4O2S
InChi Key: JDAJYNHGBUXIKS-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C(Cl)=C1Cl)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)