accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 347389-74-6

IUPAC Name: 2-ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 152
Molecular Formula: C8H13BO2
InChi Key: ZEOXJIIUMORBNN-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C#C
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results