accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34756-16-6

IUPAC Name: 3-(5-methylfuran-2-yl)propanal
Synonyms: 3-(5-methylfuran-2-yl)propanal
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: WPBUABKBDHGOAJ-UHFFFAOYSA-N
SMILES: CC1=CC=C(CCC=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results