CAS RN 34767-44-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Cyclopentadienylzirconium trichloride, 98+%, Thermo Scientific Chemicals
CAS: 34767-44-7 MDL Number: MFCD00015756
2
Cyclopentadienylzirconium(IV) Trichloride 96.0+%, TCI America™
CAS: 34767-44-7 Molecular Formula: C5H5Cl3Zr Synonym: Trichloro(cyclopentadienyl)zirconium(IV)