accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 348-36-7

IUPAC Name: ethyl 5-fluoro-1H-indole-2-carboxylate
Synonyms: ethyl 5-fluoro-1H-indole-2-carboxylate
Molecular Weight (g/mol): 207.2
Molecular Formula: C11H10FNO2
InChi Key: VIKOQTQMWBKMNA-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC(F)=CC=C2N1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)