accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3481-02-5

IUPAC Name: cyclopropyl(phenyl)methanone
Synonyms: cyclopropyl phenyl ketone
Molecular Weight (g/mol): 146.19
Molecular Formula: C10H10O
InChi Key: PJRHFTYXYCVOSJ-UHFFFAOYSA-N
SMILES: O=C(C1CC1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)