accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNOO

CAS RN 3481-09-2

IUPAC Name: 2-chloro-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: N-chlorophthalimide
Molecular Weight (g/mol): 181.58
Molecular Formula: C8H4ClNO2
InChi Key: WDRFYIPWHMGQPN-UHFFFAOYSA-N
SMILES: ClN1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results