CAS RN 3483-12-3
IUPAC Name:
(2R,3R)-1,4-disulfanylbutane-2,3-diol
Synonyms:
L-dithiothreitol cleland reagent sputolysin meso-2,3-butanediol L-dtt clelands reagent reagent, cleland 2.3-butylene glycol (R,R)-2,3-butylene glycol reagent, cleland's
Molecular Weight (g/mol):
154.24
Molecular Formula:
C4H10O2S2
InChi Key:
VHJLVAABSRFDPM-IMJSIDKUSA-N
SMILES:
O[C@@H](CS)[C@@H](O)CS
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water
Thermo Scientific Chemicals DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water
CAS: 3483-12-3 | C4H10O2S2 | 154.24 g/mol