accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34830-11-0

IUPAC Name: europium(3+) tris(4,7,7-trimethyl-3-oxo-2-(2,2,2-trifluoroacetyl)bicyclo[2.2.1]heptan-2-ide)
Synonyms: europium(3+) tris(4,7,7-trimethyl-3-oxo-2-(2,2,2-trifluoroacetyl)bicyclo[2.2.1]heptan-2-ide)
Molecular Weight (g/mol): 893.68
Molecular Formula: C36H42EuF9O6
InChi Key: VJBOGQMNBRGINA-UHFFFAOYNA-N
SMILES: [Eu+3].CC1(C)C2CCC1(C)C(=O)[C-]2C(=O)C(F)(F)F.CC1(C)C2CCC1(C)C(=O)[C-]2C(=O)C(F)(F)F.CC1(C)C2CCC1(C)C(=O)[C-]2C(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results