accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34841-09-3

IUPAC Name: 3-amino-3-(3,4-dimethoxyphenyl)propanoic acid
Synonyms: 3-amino-3-(3,4-dimethoxyphenyl)propanoic acid
Molecular Weight (g/mol): 225.24
Molecular Formula: C11H15NO4
InChi Key: FGCXSFRGPCUBPW-UHFFFAOYNA-N
SMILES: COC1=C(OC)C=C(C=C1)C(N)CC(O)=O
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)