accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34841-11-7

IUPAC Name: methyl 2-methoxy-5-nitrobenzoate
Synonyms: methyl 2-methoxy-5-nitrobenzoate
Molecular Weight (g/mol): 211.17
Molecular Formula: C9H9NO5
InChi Key: DOBFJVVTBNTGCW-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(OC)C=CC(=C1)[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)