accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34846-44-1

BrS

CAS RN 34846-44-1

IUPAC Name: 3-(bromomethyl)thiophene
Synonyms: 3-(bromomethyl)thiophene
Molecular Weight (g/mol): 177.06
Molecular Formula: C5H5BrS
InChi Key: KBWHYRUAHXHHFO-UHFFFAOYSA-N
SMILES: BrCC1=CSC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results