accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34846-90-7

IUPAC Name: methyl 3-methoxyprop-2-enoate
Synonyms: methyl 3-methoxyprop-2-enoate
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: AUTCCPQKLPMHDN-UHFFFAOYSA-N
SMILES: COC=CC(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results