CAS RN 3485-62-9
CAS RN 3485-62-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Clidinium bromide, MilliporeSigma™ Supelco™
Clidinium bromide is an anticholinergic, antisecretory and antispasmodic drug, widely used in treating anxiety-related conditions and spastic colon by inhibiting the action of parasympathetic innervations thus decreasing the secretions of stomach acid and slowing the intestines.