accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3485-84-5

IUPAC Name: 2-ethenyl-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-ethenylisoindole-1,3-dione
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: IGDLZDCWMRPMGL-UHFFFAOYSA-N
SMILES: C=CN1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results