accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34880-70-1

CH3OH3COSiCH3H3CCH3(E/Z)

CAS RN 34880-70-1

IUPAC Name: [(1-methoxyprop-1-en-1-yl)oxy]trimethylsilane
Synonyms: [(1-methoxyprop-1-en-1-yl)oxy]trimethylsilane
Molecular Weight (g/mol): 160.29
Molecular Formula: C7H16O2Si
InChi Key: RTHAKVSHSCPUGI-UHFFFAOYSA-N
SMILES: COC(O[Si](C)(C)C)=CC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results