accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH

CAS RN 34885-03-5

IUPAC Name: (4-methylcyclohexyl)methanol
Synonyms: 4-methylcyclohexanemethanol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: OSINZLLLLCUKJH-UHFFFAOYSA-N
SMILES: CC1CCC(CO)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results