accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3489-28-9

IUPAC Name: nonane-1,9-dithiol
Synonyms: nonane-1,9-dithiol
Molecular Weight (g/mol): 192.38
Molecular Formula: C9H20S2
InChi Key: GJRCLMJHPWCJEI-UHFFFAOYSA-N
SMILES: SCCCCCCCCCS
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)