accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 349-03-1

IUPAC Name: 1-bromo-2-nitro-4-(trifluoromethyl)benzene
Synonyms: 1-bromo-2-nitro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 270.01
Molecular Formula: C7H3BrF3NO2
InChi Key: PESPBNYBZVIGRO-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1Br)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results