accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 349-43-9

IUPAC Name: ethyl 2-fluoropropanoate
Synonyms: ethyl 2-fluoropropanoate
Molecular Weight (g/mol): 120.12
Molecular Formula: C5H9FO2
InChi Key: ODMITNOQNBVSQG-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results