accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOSFOO

CAS RN 349-96-2

IUPAC Name: 4-nitrobenzene-1-sulfonyl fluoride
Synonyms: 4-nitrobenzenesulfonyl fluoride
Molecular Weight (g/mol): 205.16
Molecular Formula: C6H4FNO4S
InChi Key: HRLAPUHJWRZEIC-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)S(F)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results