accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHFFF

CAS RN 349-97-3

IUPAC Name: N-[4-(trifluoromethyl)phenyl]acetamide
Synonyms: N-[4-(trifluoromethyl)phenyl]acetamide
Molecular Weight (g/mol): 203.16
Molecular Formula: C9H8F3NO
InChi Key: DFDHFECLWHHELH-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results