accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34904-98-8

IUPAC Name: [2-(phenoxymethyl)phenyl]methanol
Synonyms: [2-(phenoxymethyl)phenyl]methanol
Molecular Weight (g/mol): 214.26
Molecular Formula: C14H14O2
InChi Key: RMRTWBZKENSMCI-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1COC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results