accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOCl

CAS RN 349089-42-5

IUPAC Name: 2-chloro-N,N-dicyclohexylbenzamide
Synonyms: 2-chloro-N,N-dicyclohexylbenzamide
Molecular Weight (g/mol): 319.87
Molecular Formula: C19H26ClNO
InChi Key: WHSMKKIHACOWQV-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C(=O)N(C1CCCCC1)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results