accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ON

CAS RN 349090-11-5

IUPAC Name: N,N-dicyclohexyl-3-methylbenzamide
Synonyms: N,N-dicyclohexyl-3-methylbenzamide
Molecular Weight (g/mol): 299.46
Molecular Formula: C20H29NO
InChi Key: YXPMPPOIMVVDFJ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C(=O)N(C1CCCCC1)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results