CAS RN 3491-12-1
IUPAC Name:
4-[4-(4-aminophenoxy)phenoxy]aniline
Synonyms:
4-[4-(4-aminophenoxy)phenoxy]aniline
Molecular Weight (g/mol):
292.34
Molecular Formula:
C18H16N2O2
InChi Key:
JCRRFJIVUPSNTA-UHFFFAOYSA-N
SMILES:
NC1=CC=C(OC2=CC=C(OC3=CC=C(N)C=C3)C=C2)C=C1
1
–
1
of
1
results
1
1,4-Bis(4-aminophenoxy)benzene 98.0+%, TCI America™
CAS: 3491-12-1 Molecular Formula: C18H16N2O2 Molecular Weight (g/mol): 292.34 MDL Number: MFCD00039153 InChI Key: JCRRFJIVUPSNTA-UHFFFAOYSA-N Synonym: Hydroquinone Bis(4-aminophenyl) Ether PubChem CID: 19012 IUPAC Name: 4-[4-(4-aminophenoxy)phenoxy]aniline SMILES: C1=CC(=CC=C1N)OC2=CC=C(C=C2)OC3=CC=C(C=C3)N